Skip to Content
Merck
All Photos(1)

Key Documents

SML1159

Sigma-Aldrich

XIB4035

≥98% (HPLC)

Synonym(s):

N4-[7-Chloro-2-[(1E)-2-(2-chlorophenyl)ethenyl]-4-quinolinyl]-N1,N1-diethyl-1,4-pentanediamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$167.00
25 MG
$661.00

$167.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$167.00
25 MG
$661.00

About This Item

Empirical Formula (Hill Notation):
C26H31Cl2N3
CAS Number:
Molecular Weight:
456.45
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$167.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=CC=C1/C=C/C2=NC3=C(C=CC(Cl)=C3)C(NC(C)CCCN(CC)CC)=C2

InChI

1S/C26H31Cl2N3/c1-4-31(5-2)16-8-9-19(3)29-26-18-22(14-12-20-10-6-7-11-24(20)28)30-25-17-21(27)13-15-23(25)26/h6-7,10-15,17-19H,4-5,8-9,16H2,1-3H3,(H,29,30)/b14-12+

InChI key

CMLVHSVFSYNMGM-WYMLVPIESA-N

Biochem/physiol Actions

XIB4035 is a reported agonist of the glial cell-line derived neurotrophic factor (GDNF) receptor GFRα1.
XIB4035 is a reported agonist of the glial cell-line derived neurotrophic factor (GDNF) receptor GFRα1. XIB4035 induces GFRα1/RET phosphorylation in Neuro2A cells.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean Jmaeff et al.
Molecular pharmacology, 98(1), 1-12 (2020-05-05)
Glial cell line-derived neurotrophic factor (GDNF) binds the GFRα1 receptor, and the GDNF-GFRα1 complex binds to and activates the transmembrane RET tyrosine kinase to signal through intracellular Akt/Erk pathways. To dissect the GDNF-GFRα1-RET signaling complex, agents that bind and activate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service