Skip to Content
Merck
All Photos(1)

Key Documents

SML2983

Sigma-Aldrich

Vandetanib

≥98% (HPLC)

Synonym(s):

N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-4-quinazolinamine, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine, ZD6474

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$141.00
25 MG
$573.00

$141.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$141.00
25 MG
$573.00

About This Item

Empirical Formula (Hill Notation):
C22H24BrFN4O2
CAS Number:
Molecular Weight:
475.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$141.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)Br)Nc2ncnc3c2cc(c(c3)OCC4CCN(CC4)C)OC

InChI

1S/C22H24BrFN4O2/c1-28-7-5-14(6-8-28)12-30-21-11-19-16(10-20(21)29-2)22(26-13-25-19)27-18-4-3-15(23)9-17(18)24/h3-4,9-11,13-14H,5-8,12H2,1-2H3,(H,25,26,27)

InChI key

UHTHHESEBZOYNR-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, selective inhibitor of VEGF receptor kinase activity; also inhibits EGFR and RET tyrosine kinases
Vandetanib is an orally active and potent inhibitor of vascular endothelial growth factor receptor receptor 2 (VEGFR2) kinase activity that is used to treat medullary thyroid cancer. Vandetanib also inhibits epidermal growth factor receptor (EGFR), and RET tyrosine kinases.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2

Target Organs

Gastrointestinal tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent F Hennequin et al.
Journal of medicinal chemistry, 45(6), 1300-1312 (2002-03-08)
We have previously shown that 4-anilinoquinazolines can be potent inhibitors of vascular endothelial growth factor (VEGF) receptor (Flt-1 and KDR) tyrosine kinase activity. A novel subseries of 4-anilinoquinazolines that possess basic side chains at the C-7 position of the quinazoline
Ole Vincent Ancker et al.
International journal of molecular sciences, 21(1) (2019-12-22)
Thyroid cancer is the most common endocrine malignancy. Most thyroid cancer types respond well to conventional treatment consisting of surgery and radioactive iodine (RAI) therapy. Unfortunately, some thyroid cancer types are resistant to surgical and RAI therapy. Multikinase inhibitors (MKIs)
Francesca Carlomagno et al.
Cancer research, 62(24), 7284-7290 (2002-12-25)
RET/papillary thyroid carcinoma (PTC) oncogenes, generated by recombination of the tyrosine kinase-encoding domain of RET with different heterologous genes, are prevalent in papillary carcinomas of the thyroid. Point mutations of RET cause multiple endocrine neoplasia type 2 (MEN2) familial cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service