Skip to Content
Merck
All Photos(1)

Key Documents

SML3195

Sigma-Aldrich

AZD0530

≥98% (HPLC)

Synonym(s):

4-(6-Chloro-2,3-methylenedioxyanilino)-7-[2-(4-methylpiperazin-1-yl)ethoxy]-5-tetrahydropyran-4-yloxyquinazoline, 7-[2-[4-Methylpiperazin-1-yl]ethoxy]-5-[[tetrahydropyran-4-yl]oxy]-4-[[6-chloro-2,3-methylenedioxyphenyl]amino]quinazoline, AZD 0530, AZD-0530, N-(5-Chloro-1,3-benzodioxol-4-yl)-7-[2-(4-methyl-1-piperazinyl)ethoxy]-5-[(tetrahydro-2H-pyran-4-yl)oxy]-4-quinazolinamine, Saracatinib

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
10 MG
$518.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
10 MG
$518.00

About This Item

Empirical Formula (Hill Notation):
C27H32ClN5O5
CAS Number:
Molecular Weight:
542.03
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(c6c(cc1)OCO6)Nc2ncnc3c2c(cc(c3)OCCN5CCN(CC5)C)OC4CCOCC4

InChI

1S/C27H32ClN5O5/c1-32-6-8-33(9-7-32)10-13-35-19-14-21-24(23(15-19)38-18-4-11-34-12-5-18)27(30-16-29-21)31-25-20(28)2-3-22-26(25)37-17-36-22/h2-3,14-16,18H,4-13,17H2,1H3,(H,29,30,31)

InChI key

OUKYUETWWIPKQR-UHFFFAOYSA-N

Biochem/physiol Actions

AZD0530 is an orally active, potent tyrosine kinase inhibitor against Src family kinases (IC50 = 2.7 nM/c-Src, <4 nM/Lck, 4 nM/c-Yes, 5 nM/Lyn, 10 nM/Fyn, 10 nM/Fgr, 11 nM/Blk), v-Abl (IC50 = 30 nM) and EGFR (L861Q/L858R/wt IC50 = 4/5/66 nM). AZD0530 exhibits anti-proliferation activity in cancer cultures (IC50 = 80-700 nM; Src Y530F NIH 3T3, LoVo, K562, PC-9, SW 403, PC-3) and shows efficacy against several human tumor xenograft growth in mice in vivo (6.25-50 mg/kg/day p.o.; MDA-MB-231, AsPc-1, Calu-6, BT474C).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service