Skip to Content
Merck
All Photos(1)

Key Documents

SML3104

Sigma-Aldrich

Pazopanib

≥98% (HPLC)

Synonym(s):

5-[[4-[(2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino]pyrimidin-2-yl]amino]-2-methylbenzenesulfonamide, 5-[[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]-2-pyrimidinyl]amino]-2-methylbenzenesulfonamide, GW 786034, GW786034

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$129.00
50 MG
$518.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$129.00
50 MG
$518.00

About This Item

Empirical Formula (Hill Notation):
C21H23N7O2S
CAS Number:
Molecular Weight:
437.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1N=C2C=C(N(C3=NC(NC4=CC=C(C(S(N)(=O)=O)=C4)C)=NC=C3)C)C=CC2=C1C

InChI

1S/C21H23N7O2S/c1-13-5-6-15(11-19(13)31(22,29)30)24-21-23-10-9-20(25-21)27(3)16-7-8-17-14(2)28(4)26-18(17)12-16/h5-12H,1-4H3,(H2,22,29,30)(H,23,24,25)

InChI key

CUIHSIWYWATEQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pazopanib is an orally active receptor tyrosine kinases inhibitor against VEGFR1/2/3 (IC50 = 10/30/47 nM), PDGFR1/2 (IC50 = 84/71 nM) and c-Kit (IC50 = 74 nM), exhibiting reduced or no potency toward FGFR1/3 (IC50 = 140/130 nM), c-fms (IC50 = 146 nM), and other kinases tested (IC50 from >400 nM to >20 μM). Pazopanib exhibits anti-cancer efficacy in cultures (IC50 from 5 to 15 μg/mL; multiple myeloma migration and survival) and in mice in vivo (30-100 mg/kg/day p.o. against human MM xenografts) via angiogenesis inhibition and apoptosis induction.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B - STOT RE 2

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service