Skip to Content
Merck
All Photos(1)

Key Documents

SML3043

Sigma-Aldrich

XL188

≥98% (HPLC)

Synonym(s):

((R)-N-(3-((4-hydroxy-1-(3-phenylbutanoyl)piperidin-4-yl)methyl)-4-oxo-3,4-dihydroquinazolin-7-yl)-3-(4-methylpiperazin-1-yl)propanamide, N-[3,4-Dihydro-3-[[4-hydroxy-1-[(3R)-1-oxo-3-phenylbutyl]-4-piperidinyl]methyl]-4-oxo-7-quinazolinyl]-4-methyl-1-piperazinepropanamide, XL-188

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$159.00
25 MG
$639.00

$159.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$159.00
25 MG
$639.00

About This Item

Empirical Formula (Hill Notation):
C32H42N6O4
CAS Number:
Molecular Weight:
574.71
UNSPSC Code:
12352200
NACRES:
NA.77

$159.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(CC2(O)CCN(C(C[C@@H](C)C3=CC=CC=C3)=O)CC2)C=NC4=CC(NC(CCN5CCN(C)CC5)=O)=CC=C41

InChI

1S/C32H42N6O4/c1-24(25-6-4-3-5-7-25)20-30(40)37-14-11-32(42,12-15-37)22-38-23-33-28-21-26(8-9-27(28)31(38)41)34-29(39)10-13-36-18-16-35(2)17-19-36/h3-9,21,23-24,42H,10-20,22H2,1-2H3,(H,34,39)/t24-/m1/s1

InChI key

QLBYDWATOPNXBG-XMMPIXPASA-N

Biochem/physiol Actions

Highly selective and potent (IC50 90 nM) non covalent inhibitor of USP7
XL188 is a highly selective and potent (IC50 90 nM) non covalent inhibitor of USP7 that binds to the USP7 active site. XL188 facilitates degradation of HDM2, and increases the levels of p53 and p21 in MCF7 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Lamberto et al.
Cell chemical biology, 24(12), 1490-1500 (2017-10-24)
Deubiquitinating enzymes (DUBs) have garnered significant attention as drug targets in the last 5-10 years. The excitement stems in large part from the powerful ability of DUB inhibitors to promote degradation of oncogenic proteins, especially proteins that are challenging to
Björn Stolte et al.
The Journal of experimental medicine, 215(8), 2137-2155 (2018-07-27)
Ewing sarcoma is a pediatric cancer driven by EWS-ETS transcription factor fusion oncoproteins in an otherwise stable genomic background. The majority of tumors express wild-type TP53, and thus, therapies targeting the p53 pathway would benefit most patients. To discover targets
Jonathan W Bushman et al.
Cell chemical biology, 28(1), 78-87 (2020-10-03)
Deubiquitinating enzymes (DUBs) catalyze the removal of ubiquitin, thereby reversing the activity of E3 ubiquitin ligases and are central to the control of protein abundance and function. Despite the growing interest in DUBs as therapeutic targets, cellular functions for DUBs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service