Skip to Content
Merck
All Photos(2)

Documents

143367

Sigma-Aldrich

1-Adamantyl isothiocyanate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NS
CAS Number:
Molecular Weight:
193.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

166-168 °C (lit.)

SMILES string

S=C=NC12CC3CC(CC(C3)C1)C2

InChI

1S/C11H15NS/c13-7-12-11-4-8-1-9(5-11)3-10(2-8)6-11/h8-10H,1-6H2/t8-,9+,10-,11-

InChI key

YPKFLUARLJRPQM-BIBSGERRSA-N

General description

1-Adamantyl isothiocyanate undergoes nucleophilic addition reaction with pyrrolidine, piperidine, 3-hydroxypiperidine and 4-hydroxypiperidine to yield N′,N′-disubstituted N-(1-adamantyl)-thiourea derivatives.

Application

1-Adamantyl isothiocyanate was used in the synthesis of:
  • tris(2-mercapto-1-adamantylimidazolyl)hydroborato ligand
  • 1-adamantyl-3-(2-(7-methoxy-1,2,3,4-tetrahydroacridin-9-ylamino)alkane)thiourea 2,3-dihydroxysuccinates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Kreutzberger et al.
Arzneimittel-Forschung, 27(5), 969-972 (1977-01-01)
By nucleophilic addition of pyrrolidine (2a), piperidine (2b), 3-hydroxypiperidine (2c), and 4-hydroxypiperidine (2d) on 1-adamantyl-isothiocyanate (1), the N',N'-disubstituted N-(1-adamantyl)-thiourea derivatives 3a, 3b, 3c, and 3d are accessible. Particularly 3d exhibits remarkable antiviral activity vaccinia and herpes virus.
Van Hien Pham et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-17)
Reaction of 4-(1-adamantyl)-3-thiosemicarbazide (1) with numerous substituted acetophenones and benzaldehydes yielded the corresponding thiosemicarbazones containing adamantane skeletons. The synthesized compounds were evaluated for their in vitro activities against some Gram-positive and Gram-negative bacteria, and the fungus Candida albicans, and cytotoxicity
Kevin Yurkerwich et al.
Inorganic chemistry, 50(24), 12284-12295 (2011-09-29)
The tris(2-mercapto-1-adamantylimidazolyl)hydroborato ligand, [Tm(Ad)], has been synthesized via the reaction of 1-adamantyl-2-mercaptoimidazole with MBH(4) (M = Li, K). [Tm(Ad)]M has been used to synthesize a variety of compounds of the main-group and transition elements, including [Tm(Ad)]ZnI, {[Tm(Ad)]GaI}[GaI(4)], {[Tm(Ad)]GaCl}[GaCl(4)], {[Tm(Ad)]GaGa[Tm(Ad)]}[GaCl(4)](2), {[Tm(Ad)](2)In}[InI(4)]
Katarina Spilovska et al.
Molecules (Basel, Switzerland), 18(2), 2397-2418 (2013-02-23)
A structural series of 7-MEOTA-adamantylamine thioureas was designed, synthesized and evaluated as inhibitors of human acetylcholinesterase (hAChE) and human butyrylcholinesterase (hBChE). The compounds were prepared based on the multi-target-directed ligand strategy with different linker lengths (n = 2-8) joining the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service