Skip to Content
Merck
All Photos(1)

Documents

PZ0305

Sigma-Aldrich

WAY-208466 dihydrochloride

≥98% (HPLC)

Synonym(s):

3-[(3-Fluorophenyl)sulfonyl]-N,N-dimethyl-1H-pyrrolo[2,3-b]pyridine-1-ethanamine hydrochloride (1:2), WAY 208466 dihydrochloride, WAY208466 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18FN3O2S · 2HCl
CAS Number:
Molecular Weight:
420.33
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CN(C)CCN1C=C(S(C2=CC=CC(F)=C2)(=O)=O)C3=CC=CN=C31

InChI

1S/C17H18FN3O2S/c1-20(2)9-10-21-12-16(15-7-4-8-19-17(15)21)24(22,23)14-6-3-5-13(18)11-14/h3-8,11-12H,9-10H2,1-2H3

InChI key

SFSFIDVAEMDPIP-UHFFFAOYSA-N

Biochem/physiol Actions

WAY-208466 is a high affinity (Ki = 4.8 nM) 5-HT6 receptor agonist. In HeLa cells expressing cloned human 5-HT6, the compound induced cAMP production (EC50 = 7.3 nM; Emax = 100%). WAY-208466 increases cortical GABA levels and exhibits anxiolytic activity in rat studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service