Skip to Content
Merck
All Photos(1)

Key Documents

SML2964

Sigma-Aldrich

Gabexate mesylate

≥97% (HPLC)

Synonym(s):

Gabexate mesylate, 4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxy]-benzoic acid ethyl ester mesylate, FOY, p-Carbethoxyphenyl ε-guanidineocaproate mesylate, 4-[[6-[(Aminoiminomethyl)amino]-1-oxohexyl]oxyl]benzoic acid ethyl ester mesylate salt, Megacart

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$137.00
25 MG
$553.00

$137.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$137.00
25 MG
$553.00

About This Item

Empirical Formula (Hill Notation):
C16H23N3O4 · CH4O3S
CAS Number:
Molecular Weight:
417.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$137.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CCOC(=O)c1ccc(OC(=O)CCCCCNC(N)=N)cc1

InChI

1S/C16H23N3O4.CH4O3S/c1-2-22-15(21)12-7-9-13(10-8-12)23-14(20)6-4-3-5-11-19-16(17)18;1-5(2,3)4/h7-10H,2-6,11H2,1H3,(H4,17,18,19);1H3,(H,2,3,4)

InChI key

DNTNDFLIKUKKOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gabexate mesylate is a competitive inhibitor of serine proteases that displays high selectivity for TMPRSS2 (Transmembrane protease serine 2) (IC50 = 130 nM) with little or no activity towards several MMPs, caspases and USPs (>> 10 μM). Gabexate mesylate was reported to decrease inflammatory cytokines production and be useful as an anticoagulant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehtap Yuksel et al.
The Journal of pharmacology and experimental therapeutics, 305(1), 298-305 (2003-03-22)
Gabexate mesilate, a synthetic protease inhibitor, was shown to be effective in treating patients with sepsis-associated disseminated intravascular coagulation in which tumor necrosis factor-alpha (TNF-alpha) plays a critical role. We demonstrated that gabexate mesilate reduced lipopolysaccharide (LPS)-induced tissue injury by
Mizuki Yamamoto et al.
Viruses, 12(6) (2020-06-14)
Although infection by SARS-CoV-2, the causative agent of coronavirus pneumonia disease (COVID-19), is spreading rapidly worldwide, no drug has been shown to be sufficiently effective for treating COVID-19. We previously found that nafamostat mesylate, an existing drug used for disseminated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service