Skip to Content
Merck
All Photos(1)

Key Documents

420099

Millipore

JAK Inhibitor I

≥98% (HPLC), solid, JAK inhibitor, Calbiochem®

Synonym(s):

JAK Inhibitor I, 2-(1,1-Dimethylethyl)-9-fluoro-3,6-dihydro-7H-benz[h]-imidaz[4,5-f]isoquinolin-7-one, Pyridone 6, P6, DBI, JAK1 Inhibitor I, JAK2 Inhibitor I, JAK3 Inhibitor X

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$273.00
1 MG
$493.00

$273.00


Estimated to ship on28 May 2025



Select a Size

Change View
500 μG
$273.00
1 MG
$493.00

About This Item

Empirical Formula (Hill Notation):
C18H16FN3O
CAS Number:
Molecular Weight:
309.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$273.00


Estimated to ship on28 May 2025


Product Name

JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC₅₀ = 15 nM), JAK2 (IC₅₀ = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC₅₀ = 1 nM).

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1cc2c(c3[nH]c(nc3c4c2c(ncc4)O)C(C)(C)C)cc1

InChI

1S/C18H16FN3O/c1-18(2,3)17-21-14-10-5-4-9(19)8-12(10)13-11(15(14)22-17)6-7-20-16(13)23/h4-8H,1-3H3,(H,20,23)(H,21,22)

InChI key

VNDWQCSOSCCWIP-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, and ATP-competitive inhibitor of Janus protein tyrosine kinases (JAKs). Displays potent inhibitory activity against JAK1 (IC50 = 15 nM for murine JAK1), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM), and Tyk2 (IC50 = 1 nM). Inhibits other kinases at much higher concentrations. Shown to inhibit IL2- and IL4-dependent proliferation of CTLL cells and block the phosphorylation of STAT5, and further induce growth inhibition of multiple myeloma cells expressing activated JAKs and STAT3, unlike AG 490 (Cat. No. 658401).
A potent, reversible, cell-permeable, and ATP-competitive inhibitor of Janus protein tyrosine kinases (JAKs). Displays potent inhibitory activity against JAK1 (IC50 = 15 nM for murine JAK1), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM), and Tyk2 (IC50 = 1 nM). Inhibits other kinases at much higher concentrations. Shown to inhibit IL2- and IL4-dependent proliferation of CTLL cells and block the phosphorylation of STAT5; and further induce growth inhibition of multiple myeloma cells expressing activated JAKs and STAT3, unlike AG 490 (Cat. No. 658401). A 10 mM (500 µg/162 µl) solution of JAK Inhibitor I (Cat. No. 420097) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
murine JAK1
Product competes with ATP.
Reversible: yes
Target IC50: 15 nM against murine JAK1; 1 nM against JAK2; 1 nM against Tyk2
Target Ki: 5 nM against JAK3

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Pedranzini, L., et al. 2006. Cancer Res.66, 9714.
Lucet, I.S., et al. 2005. Blood107, 176.
Thompson, J.E., et al. 2002. Bioorg. Med. Chem. Lett.12, 1219.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pratap Karki et al.
Cellular signalling, 53, 246-255 (2018-10-20)
Exposure to particulate matter (PM) associated with air pollution remains a major public health concern, as it has been linked to significant increase in cardiopulmonary morbidity and mortality. Lung endothelial cell (EC) dysfunction is one of the hallmarks of cardiovascular
Dongbum Kim et al.
BMB reports, 54(8), 425-430 (2021-04-10)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) induces coronavirus disease 2019 (COVID-19) and may increase the risk of adverse outcomes in lung cancer patients. In this study, we investigated the expression and function of mucin 1 (MUC1) after SARS-CoV-2 infection
Qin Hao et al.
Cells, 11(3) (2022-02-16)
Necroptosis, a form of programmed lytic cell death, has emerged as a driving factor in the pathogenesis of acute lung injury (ALI). As ALI is often associated with a cytokine storm, we determined whether pro-inflammatory cytokines modulate the susceptibility of
Chi-Shuan Fan et al.
Cells, 11(2) (2022-01-22)
M2-polarization and the tumoricidal to tumor-promoting transition are commonly observed with tumor-infiltrating macrophages after interplay with cancer cells or/and other stroma cells. Our previous study indicated that macrophage M2-polarization can be induced by extracellular HSP90α (eHSP90α) secreted from endothelial-to-mesenchymal transition-derived
Fuyuko Takata et al.
Journal of cellular biochemistry, 119(11), 9055-9063 (2018-08-05)
Oncostatin M (OSM) is a member of the interleukin (IL)-6 family cytokines. We previously demonstrated that OSM induces blood-brain barrier (BBB) impairment. However, functional characterization of IL-6 family cytokines in BBB regulation and the cytokine-related intracellular signaling pathway remain unclear.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service