Skip to Content
Merck
All Photos(1)

Key Documents

420128

Sigma-Aldrich

JNK Inhibitor II

InSolution, ≥98%

Synonym(s):

InSolution JNK Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$189.00

$189.00


Estimated to ship on24 May 2025



Select a Size

Change View
5 MG
$189.00

About This Item

Empirical Formula (Hill Notation):
C14H8N2O
Molecular Weight:
220.23
UNSPSC Code:
12352200
NACRES:
NA.77

$189.00


Estimated to ship on24 May 2025


Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow-brown

shipped in

wet ice

storage temp.

−20°C

General description

A potent, cell-permeable, selective, and reversible inhibitor of c-Jun N-terminal kinase (JNK) (IC50 = 40 nM for JNK-1 and JNK-2 and 90 nM for JNK-3). The inhibition is competitive with respect to ATP. Exhibits over 300-fold greater selectivity for JNK as compared to ERK1 and p38-2 MAP kinases. Inhibits the phosphorylation of c-Jun and blocks the expression of IL-2, IFN-γ, TNF-α, and COX-2 in cells. Blocks IL-1-induced accumulation of phospho-Jun and induction of c-Jun transcription.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JNK 1, JNK 2, JNK 3
Product competes with ATP.
Reversible: yes
Target IC50: 40 nM for JNK-1 and JNK-2 and 90 nM for JNK-3

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 50 mM (5 mg/454 µl) solution of JNK Inhibitor II (Cat. No. 420119) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C) for short-term storage or freeze (-20°C) for long-term storage.

Other Notes

Bennett, B.L., et al. 2001. Proc. Natl. Acad. Sci. USA98, 13681.
Han, Z., et al. 2001. J. Clin. Invest.108, 73.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Rong Fan et al.
iScience, 26(6), 106992-106992 (2023-06-28)
Nuclear deformation has been observed in some cancer cells for decades, but its underlying mechanism and biological significance remain elusive. To address these questions, we employed human lung cancer A549 cell line as a model in context with transforming growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service