Skip to Content
Merck
All Photos(1)

Key Documents

SML2495

Sigma-Aldrich

MLS1082

≥98% (HPLC)

Synonym(s):

2-Cyclohexyl-10-methyl-3-phenylpyrimido[4,5-b]quinoline-4,5(3H,10H)-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$133.00
25 MG
$428.00

$133.00


Check Cart for Availability


Select a Size

Change View
5 MG
$133.00
25 MG
$428.00

About This Item

Empirical Formula (Hill Notation):
C24H23N3O2
CAS Number:
Molecular Weight:
385.46
UNSPSC Code:
12352200
NACRES:
NA.77

$133.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to gray-brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C(N=C(C3CCCCC3)N(C4=CC=CC=C4)C2=O)N(C)C5=CC=CC=C51

Biochem/physiol Actions

D1 dopamine receptor PAM that binds to a different allosteric site than MLS6585 (CID 2886111)
MLS1082 is a D1 dopamine receptor positive allosteric modulator (PAM) that binds to a different allosteric site than than MLS6585 (CID 2886111). When given together, they cause an additive potentiation of dopamine potency for both cAMP accumulation and β-arrestin recruitment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service