Skip to Content
Merck
All Photos(1)

Key Documents

SML2019

Sigma-Aldrich

DS44170716

≥98% (HPLC)

Synonym(s):

(3aR,4S,9bS)-rel-8-bromo-3a,4,5,9b-tetrahydro-4-(4-methylphenyl)-3H-cyclopenta[c]quinoline-6-carboxylic acid, 8-Bromo-3a,4,5,9b-tetrahydro-4-(4-methylphenyl)-3H-cyclopenta[c]quinoline-6-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$120.00
25 MG
$606.00

$120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$120.00
25 MG
$606.00

About This Item

Empirical Formula (Hill Notation):
C20H18BrNO2
CAS Number:
Molecular Weight:
384.27
UNSPSC Code:
12352200
NACRES:
NA.77

$120.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

DS44170716 is an inhibitor of Ca2+-induced mitochondrial permeability transition. It has a different mechanism of action from cyclosporin A that is not dependent on PPIF, a peptidyl-prolyl cis-trans isomerase associated with the mitochondrial permeability transition pore (MPTP). Instead DS44170716 blocks Ca2+ flux into the mitochondria by decreasing mitochondrial membrane potential and inhibits enzyme activities of mitochondrial respiratory chain complex III, IV and V. It showed a similar potent protective effect against Ca2+-induced death in human liver cells as Cyclosporin A.
Inhibitor of Ca2+-induced mitochondrial permeability transition

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service