Skip to Content
Merck
All Photos(1)

Key Documents

SML3139

Sigma-Aldrich

ITE

≥98% (HPLC)

Synonym(s):

2-(1′H-Indole-3′-carbonyl)thiazole-4-carboxylic acid methyl ester, 2-(1H-Indole­3-carbonyl)thiazole­-4-­carboxylic acid methyl ester, Methyl 2-(1H-indole-3-carbonyl)thiazole-4-carboxylate, 2-(1’H-Indole-3′-carbonyl)-thiazole-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$129.00
25 MG
$518.00

$129.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$129.00
25 MG
$518.00

About This Item

Empirical Formula (Hill Notation):
C14H10N2O3S
CAS Number:
Molecular Weight:
286.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$129.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

COC(=O)c1csc(n1)C(=O)c2c[nH]c3ccccc23

InChI

1S/C14H10N2O3S/c1-19-14(18)11-7-20-13(16-11)12(17)9-6-15-10-5-3-2-4-8(9)10/h2-7,15H,1H3

InChI key

KDDXOGDIPZSCTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Endogenous ligand identified for aryl hydrocarbon receptor (AHR); ligand inducible transcription factor.
ITE is an endogenous aryl hydrocarbon (Ah) receptor (AhR) agonist with broad-spectrum species reactivity and 5-fold higher potency than beta-naphthoflavone (by cell-based reporter assay). ITE upregulates CYP1A1/downregulates AhR in Hepa cells (0.1-5 μM), and induces fetus AhR activation in pregnant mice in vivo (1.4mg ITE/mL DMSO/kg bw i.v. on gestation day/GD 14, fetal AhR activation assessed on GD15).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service