Skip to Content
Merck
All Photos(1)

Key Documents

SML1481

Sigma-Aldrich

FzM1

≥98% (HPLC)

Synonym(s):

1-(3-Hydroxy-5-(thiophen-2-yl)phenyl)-3-(naphthalen-2-yl)urea

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$700.00

$700.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$700.00

About This Item

Empirical Formula (Hill Notation):
C21H16N2O2S
CAS Number:
Molecular Weight:
360.43
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$700.00


Estimated to ship on01 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C=CC=C2)=C2C=C1)NC3=CC(C4=CC=CS4)=CC(O)=C3

InChI

1S/C21H16N2O2S/c24-19-12-16(20-6-3-9-26-20)11-18(13-19)23-21(25)22-17-8-7-14-4-1-2-5-15(14)10-17/h1-13,24H,(H2,22,23,25)

InChI key

OBTSACXGWYGCPK-UHFFFAOYSA-N

Biochem/physiol Actions

FzM1 is an allosteric ligand of the Frizzled4 (Fz4) receptor and a Wnt/β-catenin pathway inhibitor, found by a screen for pharmacological chaperones for a misfolded mutant of the Frizzled4 (Fz4) receptor. FzM1 is believed to induce conformational changes in Fz4 by interacting with the third intracellular loop, ICL3, inhibiting binding of dishevelled (Dsh) and hampering the formation of the Fz4-Dsh complex that is necessary for β-catenin nuclear transport and ultimately transcription of TCF/LEF-regulated genes. FzM1 was tested on two tumor cell lines. U87MG glioblastoma cells acquired a more differentiated phenotype on application. FzM1 and FzM1alk also sped up the differentiation of Caco-2 cells. FzM1 has a log EC50 value of 5.74 for inhibition of Wnt antagonism.
FzM1 is an allosteric ligand of the Frizzled4 (Fz4) receptor; Wnt/β-catenin pathway inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serena F Generoso et al.
Nature chemical biology, 11(4), 280-286 (2015-03-10)
Upon binding, ligands can chaperone their protein targets by preventing them from misfolding and aggregating. Thus, an organic molecule that works as folding chaperone for a protein might be its specific ligand, and, similarly, the chaperone potential could represent an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service