Skip to Content
Merck
All Photos(1)

Key Documents

SML3219

Sigma-Aldrich

Tenofovir alafenamide

≥98% (HPLC)

Synonym(s):

9-[(R)-2-[[(S)-[[(S)-1-(Isopropoxycarbonyl)ethyl]amino]phenoxyphosphinyl]-methoxy]-propyl]adenine, GS 7340, GS-7340, GS7340, TAF

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$129.00
50 MG
$518.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$129.00
50 MG
$518.00

About This Item

Empirical Formula (Hill Notation):
C21H29N6O5P
CAS Number:
Molecular Weight:
476.47
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$129.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(OC([C@@H](N[P@](CO[C@@H](CN1C2=NC=NC(N)=C2N=C1)C)(OC3=CC=CC=C3)=O)C)=O)C

InChI

1S/C21H29N6O5P/c1-14(2)31-21(28)16(4)26-33(29,32-17-8-6-5-7-9-17)13-30-15(3)10-27-12-25-18-19(22)23-11-24-20(18)27/h5-9,11-12,14-16H,10,13H2,1-4H3,(H,26,29)(H2,22,23,24)/t15-,16+,33+/m1/s1

InChI key

LDEKQSIMHVQZJK-CAQYMETFSA-N

Biochem/physiol Actions

HIV and HBV reverse transcriptase inhibitor tenofovir isopropylalaninyl phenyl ester with greater antiviral efficacy than tenofovir disoproxil fumarate.
Tenofovir alafenamide (TAF; GS-7340) is an isopropylalaninyl phenyl ester prodrug of the nucleotide analog reverse transcriptase inhibitor (nRTI) tenofovir (TFV) against human immunodeficiency virus (HIV) and hepatitis B virus (HBV). GS-7340 more efficiently delivers TFV into lymphoid cells and tissues than tenofovir disoproxil fumarate (TDF), resulting in greater antiviral efficacy against HIV.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service