Skip to Content
Merck
All Photos(1)

Key Documents

SML1426

Sigma-Aldrich

Gap19 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

KQIEIKKFK trifluoroacetate salt, L-lLysyl-L-glutaminyl-L-isoleucyl-L-α-glutamyl-L-isoleucyl-L-lysyl-L-lysyl-L-phenylalanyl--L-lysine, Lys-Gln-Ile-Glu-Ile-Lys-Lys-Phe-Ly trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$310.00

$310.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$310.00

About This Item

Empirical Formula (Hill Notation):
C55H96N14O13 · xC2HF3O2
CAS Number:
Molecular Weight:
1161.44 (free base basis)
UNSPSC Code:
12352202
NACRES:
NA.77

$310.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

N([C@@H]([C@H](CC)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCCCN)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CCCCN)CCC(=O)N)[C@H](CC)C)CCC(=O)O

InChI

1S/C55H96N14O13/c1-5-33(3)45(69-51(77)40(25-27-44(71)72)65-54(80)46(34(4)6-2)68-50(76)39(24-26-43(61)70)62-47(73)36(60)20-10-14-28-56)53(79)64-38(22-12-16-30-58)48(74)63-37(21-11-15-29-57)49(75)67-42(32-35-18-8-7-9-19-35)52(78)66-41(55(81)82)23-13-17-31-59/h7-9,18-19,33-34,36-42,45-46H,5-6,10-17,20-32,56-60H2,1-4H3,(H2,61,70)(H,62,73)(H,63,74)(H,64,79)(H,65,80)(H,66,78)(H,67,75)(H,68,76)(H,69,77)(H,71,72)(H,81,82)/t33-,34-,36-,37-,38-,39-,40-,41-,42-,45-,46-/m0/s1

InChI key

IEAKEKFIXUZWEH-PKMKMBMKSA-N

Application

Gap19 trifluoroacetate salt has been used to inhibit Ca2+/calmodulin (CaM) gating response of connexin 43 (Cx43) gap junctions.[1]

Biochem/physiol Actions

Gap19, nonapeptide from the cytoplasmic loop of Connexin-43, is a selective blocker of connexin 43 (Cx43) hemichannel that does not block GJ or Cx40/Panx-1 channels in cardiomyocytes and astrocytes. Gap19 prevents Cx43 hemichannel opening and protect against myocardial ischemia/reperfusion injury.
Gap19, nonapeptide from the cytoplasmic loop of Connexin-43, is a selective blocker of connexin 43 (Cx43) hemichannel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium-Calmodulin Gating of Connexin43 Gap Junctions in the Absence of the pH Gating Domain.
Veenstra R D.
bioRxiv, 311613-311613 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service