Skip to Content
Merck
All Photos(1)

Key Documents

SML2533

Sigma-Aldrich

Jedi1

≥98% (HPLC)

Synonym(s):

2-Methyl-5-phenyl-3-furancarboxylic acid, 2-Methyl-5-phenyl-3-furoic acid, 2-Methyl-5-phenylfuran-3-carboxylic acid, 5-Phenyl-2-methyl-3-furancarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$178.00
25 MG
$573.00

$178.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$178.00
25 MG
$573.00

About This Item

Empirical Formula (Hill Notation):
C12H10O3
CAS Number:
Molecular Weight:
202.21
UNSPSC Code:
12352302
NACRES:
NA.77

$178.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[O-]C(=O)c1c([o]c(c1)c2ccccc2)C

InChI

1S/C12H10O3/c1-8-10(12(13)14)7-11(15-8)9-5-3-2-4-6-9/h2-7H,1H3,(H,13,14)/p-1

InChI key

VLMNACSEESRUAK-UHFFFAOYSA-M

Biochem/physiol Actions

Jedi1 is an activator of the mechanosensitive Piezo1 channel that binds at a different place from Yoda1. It exhibits long-range allosteric gating, activating Piezo1 through the extracellular side of its peripheral blade-like structure rather than the C-terminal extracellular domain of the pore.
Piezo1 channel activator

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service