Skip to Content
Merck
All Photos(1)

Key Documents

SML1412

Sigma-Aldrich

KRIBB11 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

2,6-Pyridinediamine trifluoroacetate salt, N2-(1H-Indazol-5-yl)-N6-methyl-3-nitropyridine-2,6-diamine trifluoroacetate salt, N2-1H-Indazol-5-yl-N6-methyl-3-nitro-2,6-pyridinediamine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$123.00
25 MG
$490.00

$123.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$123.00
25 MG
$490.00

About This Item

Empirical Formula (Hill Notation):
C13H12N6O2 · C2HF3O2
CAS Number:
Molecular Weight:
398.30
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$123.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNC1=NC(NC2=CC=C(NN=C3)C3=C2)=C([N+]([O-])=O)C=C1.FC(F)(C(O)=O)F

InChI

1S/C13H12N6O2.C2HF3O2/c1-14-12-5-4-11(19(20)21)13(17-12)16-9-2-3-10-8(6-9)7-15-18-10;3-2(4,5)1(6)7/h2-7H,1H3,(H,15,18)(H2,14,16,17);(H,6,7)

InChI key

DNBQMABUWGNHGB-UHFFFAOYSA-N

Biochem/physiol Actions

KRIBB11 is an inhibitor of the transcription factor Heat Shock Factor 1 (HSF1) with an IC50 value of 1.2 μM. KRIBB11 blocks induction of HSF1 downstream target proteins such as HSP27 and HSP70 by impairing the recruitment of Positive Transcription Elongation Factor b (P-TEFb) to the HSF1 transcriptional complex, preventing HSF1-mediated gene expression KRIBB11 inhibits tumor growth in vivo and acts synergistically to increase apoptosis in cancer cells treated with the HSP90 inhibitors Geldanamycin and 17-AAG.
KRIBB11 is an inhibitor of the transcription factor Heat Shock Factor 1 (HSF1).
KRIBB11 is capable of preventing the proliferation of cancer cells.[1] It has the ability to stimulate apoptosis and can arrest the cell cycle at G2/M phase.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Ju Yoon et al.
The Journal of biological chemistry, 286(3), 1737-1747 (2010-11-17)
Heat shock factor 1 (HSF1) is the master switch for heat shock protein (HSP) expression in eukaryotes. A synthetic chemical library was screened to identify inhibitors of HSF1 using a luciferase reporter under the control of a heat shock element.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service