Skip to Content
Merck
All Photos(1)

Key Documents

5.31237

Sigma-Aldrich

IRAK4 Inhibitor, Compound 26

Synonym(s):

IRAK4 Inhibitor, Compound 26, Interleukin-1 Receptor Associated Kinase 4 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$293.00

$293.00


Estimated to ship on24 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$293.00

About This Item

Empirical Formula (Hill Notation):
C25H27N5O2
CAS Number:
Molecular Weight:
429.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$293.00


Estimated to ship on24 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

94 pM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell permeable indolo[2,3-c]quinoline derivatived compound that acts as a selective and highly potent inhibitor of interleukin-1 receptor associated kinase (IRAK4; IC50 = 94 pM). Displays much reduced potency against IRAK1 (IC50 = 65 nM) and displays high selectivity over a panel of other 27 kinases, including several involved in TLR-mediated signaling. Blocks R848-induced TNF-α and IL-6 production in primary human monocytes and inhibits LPS-induced TNF-α production in mice (100 mpk). Shows minimal inhibition of Cyp2C9, 2D6, and 3A4 and has excellent microsomal stability and desirable aqueous solubility (100 μg/ml at pH 7.4).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IRAK4
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tumey, L.N., et al. 2014 Bioorg. Med. Chem. Lett. 24, 2066.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service