Skip to Content
Merck
All Photos(1)

Key Documents

SMB00406

Sigma-Aldrich

2-Azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside

≥95% (HPLC)

Synonym(s):

β-D-GalNAc-Et-N3, 2-Azidoethyl N-acetyl-β-D-galactosaminide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$556.00

$556.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$556.00

About This Item

Empirical Formula (Hill Notation):
C10H18N4O6
CAS Number:
Molecular Weight:
290.27
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$556.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white to light yellow

storage temp.

−20°C

SMILES string

N(C(C)=O)[C@H]1[C@H](OCCN=[N+]=[N-])O[C@H](CO)[C@H](O)[C@@H]1O

InChI

1S/C11H20N4O5/c1-6(17)14-9-8(20-3-2-13-15-12)4-7(5-16)10(18)11(9)19/h7-11,16,18-19H,2-5H2,1H3,(H,14,17)/t7?,8-,9?,10+,11-/m1/s1

InChI key

XABWQXCIQZZUHY-QPGSTUFLSA-N

General description

2-azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside is an alkylated glycoside.

Biochem/physiol Actions

2-azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside is an alkylated glycoside. Alkylated glycosides are used as ligands for Click Chemistry. They may also be important as carbohydrate carriers in glycotherapeutic applications, drug carriers for site-specific delivery, drug enhancers for decreasing toxicity, and solubility and protein enhancers.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service