Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502511

Sigma-Aldrich

Anti-HECW2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

C2 and WW domain-containing protein 2, E3 ubiquitin-protein ligase HECW2, HECT, NEDD4-like E3 ubiquitin-protein ligase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$693.00

$693.00


Check Cart for Availability


Select a Size

Change View
100 μG
$693.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$693.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 175 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HECW2(57520)

General description

Anti-HECW2 Antibody detects endogenous levels of total HECW2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human HECW2.

Immunogen Range: 481-530

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vidhya Krishnamoorthy et al.
Biochimica et biophysica acta. Molecular cell research, 1865(8), 1088-1104 (2018-05-14)
Lamins constitute the major architectural proteins of the nuclear lamina that help in maintaining nuclear organization. Mutations in lamins are associated with diverse degenerative diseases, collectively termed laminopathies. HECW2, a HECT-type E3 ubiquitin ligase, is transcriptionally upregulated in HeLa cells
Richa Khanna et al.
The FEBS journal, 285(12), 2243-2262 (2018-04-21)
Lamins are key nuclear proteins which are important for maintaining nuclear structure and function. Mutations in lamins cause a spectrum of genetic diseases termed as laminopathies. RING finger containing E3 ubiquitin ligase, RNF123, is transcriptionally upregulated in cells expressing rod

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service