Skip to Content
Merck
All Photos(6)

Key Documents

HPA005980

Sigma-Aldrich

Anti-LRP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Glycoprotein 330 antibody produced in rabbit, Anti-Low-density lipoprotein receptor-related protein 2 precursor antibody produced in rabbit, Anti-Megalin antibody produced in rabbit, Anti-gp330 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GFTSMSDRPGKRCAAEGSSPLLLLPDNVRIRKYNLSSERFSEYLQDEEYIQAVDYDWDPEDIGLSVVYYTVRGEGSRFGAIKRAYIPNFESGRNNLVQEVDLKLKYVMQPDGIAVDWVGRHIYWSDVKNKRIEVAKLDGRYRKWLISTDL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRP2(4036)

Immunogen

Low-density lipoprotein receptor-related protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LRP2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Low density lipoprotein-related protein 2 is a protein encoded by the LRP2 gene in humans. It is mapped at a single site on the long arm of human chromosome 2 on the border of bands 2q24-q31. It is referred as DBS and GP330. It is a member of a family of receptors and possesses structural similarities to the low-density lipoprotein receptor. This gene is expressed by a number of specialized epithelia, including renal proximal tubules. It functions as a multi-ligand receptor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85141

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akiyoshi Nakayama et al.
Rheumatology international, 34(4), 473-476 (2013-12-25)
A recent genome-wide association study revealed that there is an association between serum uric acid (SUA) levels and rs2544390, a common variant in low-density lipoprotein-related protein 2 (LRP2/Megalin) gene. Two other variants of LRP2, rs2229268 and rs3755166, are also found
Li-Jun Ma et al.
American journal of physiology. Renal physiology, 300(5), F1203-F1213 (2011-03-04)
The mechanisms for increased risk of chronic kidney disease (CKD) in obesity remain unclear. The renin-angiotensin system is implicated in the pathogenesis of both adiposity and CKD. We investigated whether the angiotensin type 1 (AT(1)) receptor, composed of dominant AT(1a)
Ming-Tsun Tsai et al.
Clinical kidney journal, 17(4), sfae065-sfae065 (2024-04-05)
Fetuin-A is implicated in the pathogenesis of vascular calcification in chronic kidney disease (CKD); however, the relationship between fetuin-A, histopathologic lesions and long-term kidney outcomes in patients with various types of kidney disease remains unclear. We measured urinary fetuin-A levels
J R Korenberg et al.
Genomics, 22(1), 88-93 (1994-07-01)
Glycoprotein 330 (gp330) is a member of a family of receptors with structural similarities to the low-density lipoprotein receptor. Gp330 is expressed by a number of specialized epithelia, including renal proximal tubules, where it can mediate endocytosis of ligands such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service