Skip to Content
Merck
All Photos(4)

Key Documents

RDD019

Sigma-Aldrich

Potassium phosphate tribasic

anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%

Synonym(s):

Tripotassium phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$143.00
1 KG
$197.00
2.5 KG
$386.00

$143.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$143.00
1 KG
$197.00
2.5 KG
$386.00

About This Item

Linear Formula:
K3PO4
CAS Number:
Molecular Weight:
212.27
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$143.00


Available to ship on09 April 2025Details


Request a Bulk Order

grade

anhydrous
reagent grade

Quality Level

product line

Redi-Dri

Assay

≥98%

form

crystalline

quality

free-flowing

loss

≤5.0% loss on ignition

pH

11.9 (25 °C, 10 g/L)

solubility

water: 100 mg/mL, clear, colorless

density

2.564 g/mL at 25 °C (lit.)

SMILES string

[K].OP(O)(O)=O

InChI

1S/3K.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

LWIHDJKSTIGBAC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia G Flores-Hernández et al.
Polymers, 13(4) (2021-02-11)
The objective of the study was to obtain a new biodegradable graft polymer by performing two chemical processes: first, a transesterification reaction between carboxylic acid's salt and ethyl acrylate's ester, followed by polymerization of the vinyl group from the ethyl
Janaina S Lima et al.
Bioprocess and biosystems engineering, 40(4), 511-518 (2016-12-13)
Immobilization of cellulases on magnetic nanoparticles, especially magnetite nanoparticles, has been the main approach studied to make this enzyme, economically and industrially, more attractive. However, magnetite nanoparticles tend to agglomerate, are very reactive and easily oxidized in air, which has
Marta Pastor-Belda et al.
Food chemistry, 221, 76-81 (2016-12-17)
Magnetic solid phase extraction (MSPE) with cobalt ferrite nanoparticles coated with oleic acid is described for the determination of alkylphenols (APs), 4-tert-butylphenol (TBP), 4-pentylphenol (PP), 4-hexylphenol (HP), 4-tert-octylphenol (TOP), 4-n-octylphenol (OP) and 4-nonylphenol (NP) in baby foods using gas chromatography

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service