Skip to Content
Merck
All Photos(8)

Key Documents

795496

Sigma-Aldrich

Potassium phosphate dibasic

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥98%

Synonym(s):

Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$62.20
500 G
$231.00
1 KG
$362.00
2.5 KG
$503.00

$62.20


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$62.20
500 G
$231.00
1 KG
$362.00
2.5 KG
$503.00

About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98%
grade:
ACS reagent
anhydrous
form:
powder

$62.20


In StockDetails


Request a Bulk Order

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for SM 5210

product line

Redi-Dri

Assay

≥98%

form

powder

quality

free-flowing

impurities

≤0.001% Nitrogen compounds (as N)
≤0.01% Insoluble matter

loss

≤1.0% loss on drying, 105°C

pH

8.5-9.6 (25 °C, 5% in solution)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
Na: ≤0.05%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate dibasic is a useful biological buffer having a pH in the range of 8.5–9.6. Potassium phosphate is present in various forms: monobasic (KH2PO4), dibasic (KH2PO4), and tribasic (K3PO4). Potassium phosphate buffers are widely employed in equilibrium dialysis, high performance liquid chromatography (HPLC) and affinity capillary electrophoresis studies.

Application

Potassium phosphate dibasic has been used in the preparation of MMG (maltose, methyl green) medium, phosphate buffered saline and hypotonic solution.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Karimi et al.
Materials science & engineering. C, Materials for biological applications, 98, 227-240 (2019-03-01)
The aim of this study was to find an optimum dose of the synthetic amorphous calcium phosphate (ACP) nanoparticles to be incorporated in resin-modified glass ionomer cements (RMGICs) for triggering the release of PO43-/Ca2+, alkaline phosphatase (ALP) activity and osteogenic

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service