Skip to Content
Merck
All Photos(4)

Key Documents

P5629

Sigma-Aldrich

Potassium phosphate tribasic

Potassium phosphate tribasic
1 of 1 reviewers received a sample product or took part in a promotion

reagent grade, ≥98%

Synonym(s):

Tripotassium phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$38.50
500 G
$96.40
1 KG
$141.00
10 KG
$680.00

$38.50


In StockDetails


Request a Bulk Order

Select a Size

Change View
25 G
$38.50
500 G
$96.40
1 KG
$141.00
10 KG
$680.00

About This Item

Linear Formula:
K3PO4
CAS Number:
Molecular Weight:
212.27
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$38.50


In StockDetails


Request a Bulk Order

grade

reagent grade

Quality Level

Assay

≥98%

form

powder

pH

11.9 (25 °C, 10 g/L)

solubility

water: 100 mg/mL, clear, colorless

density

2.564 g/mL at 25 °C (lit.)

storage temp.

room temp

SMILES string

[K].OP(O)(O)=O

InChI

1S/3K.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

LWIHDJKSTIGBAC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate tribasic exhibits high catalytic property in trans esterification reactions.[1] It has bactericidal activity.[2]

Application

Potassium phosphate tribasic has been used:
  • in lysis buffer for the isolation of protein from Gram positive bacteria[3]
  • as a reagent for the scalable synthesis and post-modification of NU-1000 (mesoporous metal-organic framework)[4]
  • as a reagent for the plasma membrane preparation of Lilium davidii or Oryza sativa with high purity[5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bactericidal activity of tripotassium phosphate and potassium oleate on the native flora of poultry skin
Hinton Jr A and Ingram KD
Food Microbiology, 20(4), 405-410 (2003)
Tri-potassium phosphate as a solid catalyst for biodiesel production from waste cooking oil
Guan G, et al.
Fuel Processing Technology, 90(4), 520-524 (2009)
Scalable synthesis and post-modification of a mesoporous metal-organic framework called NU-1000
Wang TC, et al.
Nature Protocols, 11(1), 149-149 (2016)
Plasma Membrane Preparation from Lilium davidii and Oryza sativa Mature and Germinated Pollen
Yang N, et al.
Journal of integrative plant biology (2010)
The newly developed monoclonal antibody SA7D6 exhibits potential for detection of Staphylococcus aureus
Senevirathne A and Kim KP
Food Science and Biotechnology, 24(3), 1177-1184 (2015)

Related Content

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service