Select a Size
$1,500.00
Select a Size
About This Item
$1,500.00
Recommended Products
species reactivity
rhesus monkey
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
assay range
inter-assay cv: <10%
intra-assay cv: <12%
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rhesus macaque ... IL5(710622)
Related Categories
General description
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service