Skip to Content
Merck
All Photos(5)

Key Documents

HPA007845

Sigma-Aldrich

Anti-CASQ1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Calmitin antibody produced in rabbit, Anti-Calsequestrin, skeletal muscle isoform antibody produced in rabbit, Anti-Calsequestrin-1 precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LDFPEYDGVDRVINVNAKNYKNVFKKYEVLALLYHEPPEDDKASQRQFEMEELILELAAQVLEDKGVGFGLVDSEKDAAVAKKLGLTEVDSMYVFKGDEVIEYDGEFSADTIVEFLLDVLEDPVELIEGERELQAFENIEDEIKLIGY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASQ1(844)

General description

CASQ1 (calsequestrin 1) gene encodes a member of the calsequestrin protein family. The gene is mapped to human chromosome 1q21, a region that has been associated with type 2 diabetes in certain populations.

Immunogen

Calsequestrin-1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CASQ1 antibody produced in rabbit has been used for global protein profiling to find new molecular biomarkers for common, multifactorial disorders.
Anti-CASQ1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CASQ1 (calsequestrin 1) gene encodes a protein localized in the sarcoplasmic reticulum of fast-twitch skeletal muscle cells. It functions as a calcium storage protein regulating calcium flux via calcium channel activity control and interaction with other proteins present in the sarcoplasmic reticulum. The calcium that is released into the cytosol from the sarcoplasmic reticulum regulates the expression of GLUT4 and glucose transport in muscle. Mutations in this gene may affect insulin action on glucose uptake and glycogen synthesis in skeletal muscle and increase susceptibility to type 2 diabetes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70700

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mao Fu et al.
Diabetes, 53(12), 3292-3299 (2004-11-25)
Calsequestrin (CASQ)1 is involved in intracellular storage and release of calcium, a process that has been shown to mediate glucose transport in muscle. Its gene, CASQ1, is encoded on chromosome 1q21, a region that has been linked to type 2
Swapan Kumar Das et al.
Diabetes, 53(12), 3300-3306 (2004-11-25)
Genome-wide scans in multiple populations have identified chromosome 1q21-q24 as one susceptibility region for type 2 diabetes. To map the susceptibility genes, we first placed a dense single nucleotide polymorphism (SNP) map across the linked region. We identified two SNPs
Bernet S Kato et al.
Proteome science, 9, 73-73 (2011-11-19)
The advent of affinity-based proteomics technologies for global protein profiling provides the prospect of finding new molecular biomarkers for common, multifactorial disorders. The molecular phenotypes obtained from studies on such platforms are driven by multiple sources, including genetic, environmental, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service