Skip to Content
Merck
All Photos(1)

Key Documents

RAB0931

Sigma-Aldrich

Rhesus Macaque IL6 / Interleukin-6 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$970.00

$970.00


Check Cart for Availability


Select a Size

Change View
96 WELLS
$970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$970.00


Check Cart for Availability

species reactivity

rhesus monkey

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 40 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rhesus macaque ... IL6(705819)

General description

The IL6 ELISA kit provides for the Quantitative measurement of IL-6 in Cell Culture Supernatants, Plasma and Serum. The IL6 (interleukin-6) gene is mapped to human chromosome 7p15.3.[1] IL6 (interleukin-6) induces B-cell differentiation and is a primary regulator of inflammation. IL6 regulates tumor proliferation and inhibits apoptosis.[2] It contributes to the disease progression of coronary artery disease.[3]

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association Between the IL-6 rs1800795 Polymorphism and the Risk of Cervical Cancer: A Meta-Analysis of 1210 Cases and 1525 Controls.
Liu H, et al.
Technology in Cancer Research & Treatment, 16(5), 662-667 (2017)
The-174 G> C interleukin-6 gene polymorphism is associated with angiographic progression of coronary artery disease over a 4-year period.
Toutouzas K, et al.
Hellenic Journal of Cardiology : HJC = Hell?nik? Kardiologik? epithe?r?s?, 58(1), 80-86 (2017)
The human obesity gene map: the 2003 update.
Snyder E E, et al.
Obesity Research, 12(3), 369-439 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service