Skip to Content
Merck
All Photos(1)

Key Documents

RAB0909

Sigma-Aldrich

Rat SSSCA1 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,500.00

$1,500.00


Estimated to ship on30 May 2025



Select a Size

Change View
96 WELLS
$1,500.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,500.00


Estimated to ship on30 May 2025


species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The antibody pair provided in this kit recognizes Rat Sjogren syndrome/scleroderma autoantigen 1 / p27.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service