Skip to Content
Merck
All Photos(1)

Key Documents

RAB1136

Sigma-Aldrich

Rat Adiponectin ELISA Kit

for cell culture supernatants, plasma, and serum samples

Synonym(s):

Adiponectin Assay, Rat ELISA

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$970.00

$970.00


Check Cart for Availability


Select a Size

Change View
96 WELLS
$970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$970.00


Check Cart for Availability

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.8 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

This ELISA antibody pair detects Rat Adiponectin (ADIPOQ/ACRP30)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Adiponectin, also known as AdipoQ, is a plasma protein produced by adipocytes. Downregulation of adiponectin by mutation causes insulin resistance and type 2 diabetes.[1] The expression level of adiponectin in plasma is lower in obesity.[2][3] Adiponectin interacts with other proteins produced by adipocytes and is involved in triglyceride synthesis. Adiponectin in plasma lowers the expression of adhesion molecules in vascular endothelial cells and reduces the risk of atherogenic vascular diseases.[2]

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of adiponectin mutation with type 2 diabetes: a candidate gene for the insulin resistance syndrome
Kondo H, et al.
Diabetes, 51(7), 2325-2328 (2002)
Paradoxical decrease of an adipose-specific protein, adiponectin, in obesity
Arita Y, et al.
Biochemical and Biophysical Research Communications, 257(1), 79-83 (1999)
Genomic structure and mutations in adipose-specific gene, adiponectin
Takahashi M, et al.
International journal of obesity (2005), 24(7), 861-868 (2000)
Jóctan Pimentel Cordeiro et al.
PloS one, 17(8), e0271592-e0271592 (2022-08-12)
There are evidence that obese-resistant animals are more physically active, due to a higher rate of lipid oxidation. Efficiency in such pathways can favor greater spontaneous physical activity and, consequently, less body fat deposition. The aim of study was characterizing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service