Skip to Content
Merck
All Photos(1)

Key Documents

EZGLP1T

Millipore

Multi Species GLP-1 ELISA Kit

measures total GLP-1 levels in 20 µl - 50 µl serum, plasma or cell culture supernatent

Synonym(s):

Glucagon-like peptide-1

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$1,360.00
10 X 96 WELLS
$9,460.00

$1,360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
96 WELLS
$1,360.00
10 X 96 WELLS
$9,460.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

$1,360.00


Check Cart for Availability

Request a Bulk Order

Product Name

Multi Species GLP-1 Total ELISA, EZGLP1T-36K, This Multi Species GLP-1 Total ELISA, EZGLP1T-36K, is used to measure & quantify Glucagon Like Peptide-1 levels in Metabolism research.

Quality Level

species reactivity

human, mouse, rat

packaging

kit of 1 × 96 wells

parameter

20—50 μL sample volume (4hr assay)

assay range

linearity: 90-132%
recovery range: 90-110%
sensitivity: 1.5 pM
(MinDC)

standard curve range: 4.1-1000 pM
inter-assay cv: <12%
intra-assay cv: <5%

technique(s)

ELISA: suitable

input

sample type cell culture supernatant
sample type plasma (K2 EDTA)
sample type serum

NCBI accession no.

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... GCG(2641)

General description

Glucagon-like peptide-1 (GLP-1) is a 30 amino acid gut hormone secreted from the intestinal L-cells. The GLP-1 sequence is highly conserved among mammals and is released in its active form (7-36) in response to food intake. It is quickly degraded into its inactive form (9-36) after cleavage by DPPIV. GLP-1 reduces glucose levels by regulating pancreatic secretion, slowing gastric emptying and lowering the desire for food intake. Therefore, it is an important biomarker for the study of diabetes and obesity. This kit measures both the active (7-36) and inactive (9-36) forms of GLP-1.

Specificity

Cross Reactivty
GLP-1 is conserved in almost all mammals.
The antibody pair used in this assay is specific to GLP-1 (7-36) and (9-36) and has no cross-reactivity with GLP-2, GIP, Glucagon or Oxyntomodulin.

Application

Research Category
Metabolism
This is a 3.5-hour assay at room temperature requiring 50 uL of serum, plasma or culture samples per well.
Used to detect/quantify: Glucagon Like Peptide-1

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lindsey M Rings et al.
Journal of veterinary internal medicine, 33(6), 2752-2764 (2019-10-31)
The enteroinsular axis (EIA) comprises intestinal factors (incretins) that stimulate insulin release after PO ingestion of nutrients. Glucose-dependent insulinotropic polypeptide (GIP) and glucagon-like peptide-1 (GLP-1) are the main incretins. The EIA has not been investigated in healthy neonatal foals but
Yasutaka Takeda et al.
Physiological reports, 8(11), e14469-e14469 (2020-05-31)
The short-form glucose-dependent insulinotropic polypeptide (GIP) (1-30) is released from islet alpha cells and promotes insulin secretion in a paracrine manner in vitro. However, it is not well elucidated how GIP (1-30) is involved in glucose metabolism in vivo, since

Questions

1–3 of 3 Questions  
  1. Are there any instances of cross-reactivity to Liraglutide when testing patient samples treated with Liraglude in EZGLP1T-39K, considering that these patient samples read higher than control samples?

    1 answer
    1. The kit has not been tested for cross-reactivity to liraglutide. Since liraglutide is similar to GLP-1 7-37 and our kit has 15% cross-reactivity to GLP-1 7-37, there is a possibility of some cross-reactivity.

      Helpful?

  2. What information is available for the antibodies used in the GLP-1 Total ELISA kit (EZGLP1T-36K)?

    1 answer
    1. The sequence and epitope are proprietary. The capture involves a polyclonal rabbit anti GLP-1 total, while the detection uses a monoclonal mouse anti GLP-1 (7-36), and the immunogen is mid molecule.

      Helpful?

  3. What is the cross reactivity to other forms of GLP-1?

    1 answer
    1. The assay detects GLP-1 (1-37, 1-36, 7-37), with a moderate affinity and exhibits 12-15% cross-reactivity.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service