Skip to Content
Merck
All Photos(1)

Key Documents

RAB0504

Sigma-Aldrich

Mouse TSLP ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$915.00

$915.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$915.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$915.00


Check Cart for Availability

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1 pg/mL
standard curve range: 0.82-200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Tslp(53603)

General description

The Mouse TSLP (chemokine, interferon γ-inducible T cell α chemoattractant) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse TSLP in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse TSLP

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sha Tang et al.
Journal of cellular and molecular medicine, 24(5), 2761-2771 (2020-01-24)
Lung immune responses to respiratory pathogens and allergens are initiated in early life which will further influence the later onset of asthma. The airway epithelia form the first mechanical physical barrier to allergic stimuli and environmental pollutants, which is also
G Mócsai et al.
The British journal of dermatology, 170(3), 617-624 (2013-11-21)
Filaggrin (FLG) deficiency is a well-known predisposing factor for the development of atopic dermatitis (AD). Decreased FLG expression can be the result of haploinsufficiency or severe inflammation, which can cause acquired FLG alterations. FLG mutations are related to several clinical
S Demehri et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 44(6), 851-857 (2014-01-09)
Mouse models of atopic march suggest that systemic, skin-derived thymic stromal lymphopoietin (TSLP) mediates progression from eczema to asthma. We investigated whether circulating TSLP is associated with eczema, allergic sensitization, or recurrent wheezing in young children. A prospective analysis of
Kaori Sadakane et al.
Journal of applied toxicology : JAT, 34(8), 893-902 (2013-08-29)
Phthalate esters in plastics act as adjuvants for immunoglobulin production, which aggravates allergic disease. However, the effects of alkylphenols (used as plasticizers and surfactants) on atopic dermatitis have not been studied in detail. Therefore, the goal of the present study
Stephen F Murphy et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(9), G871-G882 (2014-09-23)
Tumor necrosis factor-induced protein 3 (TNFAIP3; also known as A20) negatively regulates NF-κB and MAPK signals to control inflammatory responses. TNFAIP3 also protects against TNF-induced cell death. Intestinal epithelial cell (IEC) expression of TNFAIP3 improves barrier function and tight junction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service