Skip to Content
Merck
All Photos(1)

Key Documents

O0884

Sigma-Aldrich

Monoclonal Anti-Oncostatin M antibody produced in mouse

clone 17001, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-OSM

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$1,740.00

$1,740.00


Check Cart for Availability


Select a Size

Change View
0.5 MG
$1,740.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

$1,740.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

17001, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: suitable
neutralization: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OSM(5008)

General description

Oncostatin M (OSM) is a growth-regulating cytokine produced by the activated T lymphocytes and macrophages. It has a pivotal role in stimulating the production of IL-6 in cultured human endothelial cells. It also facilitates the LDL receptor expression and its uptake by hepatoma cells. Further, OSM can induce synovial fibroblast-like cells to produce urokinase type plasminogen activator. Monoclonal anti-Oncostatin M antibody can be used for neutralizing the biological activity of rhOSM on the human TF-1 cell line. Mouse anti-Oncostatin M antibody reacts specifically with recombinant human OSM. The product has shown no cross-reactivity with recombinant human IL-6, IL-11, CNTF, LIF and recombinant mouse OSM.

Specificity

The antibody neutralizes the biological activity of recombinant human OSM. Does not cross-react with recombinant human IL-6, IL-11, CNTF, LIF and recombinant mouse OSM.

Immunogen

recombinant human OSM expressed in Escherichia coli.

Application

Monoclonal anti-Oncostatin M antibody can be used in western blotting and capture ELISA.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline (PBS) with 5% trehalose.

Preparation Note

Purified using protein A.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Hamilton et al.
Biochemical and biophysical research communications, 180(2), 652-659 (1991-10-31)
Cytokine regulation of synovial cell function has been considered to be involved in the pathogenesis of rheumatoid arthritis. Synoviocyte urokinase-type plasminogen activator (u-PA) expression may be relevant to the tissue remodelling, as well as to the cell migration and transformation
T J Brown et al.
Journal of immunology (Baltimore, Md. : 1950), 147(7), 2175-2180 (1991-10-01)
Endothelial cells produce immunomodulatory cytokines in response to soluble mediators of inflammatory/immune reactions. We have previously demonstrated that the leukocyte-derived cytokine, oncostatin M (Onco M) can alter endothelial cell morphology, regeneration, and fibrinolytic activity in vitro. Here we demonstrate that
R I Grove et al.
The Journal of biological chemistry, 266(27), 18194-18199 (1991-09-25)
Oncostatin M is a growth regulatory protein secreted by macrophages and activated T lymphocytes. In a hepatoma cell line (HepG2) the polypeptide very potently increased low density lipoprotein (LDL) uptake with an EC50 of 0.1-0.2 nM. The stimulation of LDL
T Kitamura et al.
Journal of cellular physiology, 140(2), 323-334 (1989-08-01)
We have established a novel cell line, designated as TF-1, from a patient with erythroleukemia, which showed complete growth dependency on granulocyte-macrophage colony-stimulating factor (GM-CSF) or on interleukin-3 (IL-3) and carried a homogeneous chromosomal abnormality (54X). Erythropoietin (EPO) also sustained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service