Skip to Content
Merck
All Photos(2)

Key Documents

RAB0469

Sigma-Aldrich

Mouse TIMP-1 ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$918.00

$918.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 KIT
$918.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

$918.00


Estimated to ship on29 May 2025


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 1.37-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Timp1(21857)

General description

The Mouse TIMP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse TIMP-1 cell lysate and tissue lysate.

Immunogen

Recombinant Mouse TIMP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirokuni Akahori et al.
Journal of hypertension, 32(7), 1534-1541 (2014-04-25)
Clinical studies have suggested the beneficial effects of statin therapy on diastolic heart failure. However, the mechanism of the beneficial effects of statin on diastolic heart failure remains unknown. We examined the effect of atorvastatin on the cardiac function of
Nejat Nizam et al.
Inflammation, 37(5), 1771-1778 (2014-05-07)
Salivary, serum matrix metalloproteinase-8 (MMP-8), tissue inhibitor of matrix metalloproteinases-1 (TIMP-1), neutrophil elastase (NE), and myeloperoxidase (MPO) levels were investigated in generalized chronic periodontitis (GCP), generalized aggressive periodontitis (GAgP), and healthy groups. Whole-mouth clinical periodontal measurements were recorded. Salivary, serum
Harpreet Sidhu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(30), 9867-9879 (2014-07-25)
Fmr1 knock-out (ko) mice display key features of fragile X syndrome (FXS), including delayed dendritic spine maturation and FXS-associated behaviors, such as poor socialization, obsessive-compulsive behavior, and hyperactivity. Here we provide conclusive evidence that matrix metalloproteinase-9 (MMP-9) is necessary to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service