Skip to Content
Merck
All Photos(2)

Key Documents

RAB0111

Sigma-Aldrich

Human Cathepsin S ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,320.00

$1,320.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$1,320.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

$1,320.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)
sample type urine

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 pg/mL
standard curve range: 4.1-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTSS(1520)

General description

The Human Cathepsin S ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Cathepsin S in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human Cathepsin S

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lili Chen et al.
Diabetes/metabolism research and reviews, 35(3), e3117-e3117 (2018-12-15)
Cathepsin S is highly expressed in subcutaneous and visceral adipose tissue. Cathepsin S correlates with central obesity and contributes to the formation and progression of atherosclerosis. Here, we sought to evaluate the association of serum cathepsin S with metabolic syndrome
Patrick Hargreaves et al.
Arthritis research & therapy, 21(1), 175-175 (2019-07-20)
Primary Sjögren syndrome (pSS) is characterized by T and B cell infiltration of exocrine glands. The cysteine protease cathepsin S (CatS) is crucially involved in MHCII processing and T cell stimulation, and elevated levels have been found in patients with
Marie Cuvelliez et al.
Scientific reports, 9(1), 19202-19202 (2019-12-18)
Heart failure (HF) remains a main cause of mortality worldwide. Risk stratification of patients with systolic chronic HF is critical to identify those who may benefit from advanced HF therapies. The aim of this study is to identify plasmatic proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service