Skip to Content
Merck
All Photos(1)

Key Documents

PZ0107

Sigma-Aldrich

CP-101537

≥98% (HPLC)

Synonym(s):

MN-2633, N-(1a,5a,6a)-3-Azabicyclo[3.1.0]hex-6-yl-carbamic acid 1,1-dimethylethyl ester, PF-3339644

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$123.00
25 MG
$487.00

$123.00


Check Cart for Availability
New, lower price on this item!


Select a Size

Change View
5 MG
$123.00
25 MG
$487.00

About This Item

Empirical Formula (Hill Notation):
C10H18N2O2
CAS Number:
Molecular Weight:
198.26
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$123.00


Check Cart for Availability
New, lower price on this item!

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N[C@H]1[C@@H]2CNC[C@H]12

InChI

1S/C10H18N2O2/c1-10(2,3)14-9(13)12-8-6-4-11-5-7(6)8/h6-8,11H,4-5H2,1-3H3,(H,12,13)/t6-,7+,8+

InChI key

QIYOMZXJQAKHEK-JIGDXULJSA-N

Gene Information

Biochem/physiol Actions

CP-101537 is a MMP inhibitor, candidate drug for myocardial infarction therapy, and antibacterial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service