Skip to Content
Merck
All Photos(1)

Documents

444278-M

Millipore

MMP-9 Inhibitor I

≥93% (HPLC), solid, MMP-9 inhibitor, Calbiochem®

Synonym(s):

MMP-9 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33N3O5S
CAS Number:
Molecular Weight:
511.63
UNSPSC Code:
12352200

product name

MMP-9 Inhibitor I, MMP-9 Inhibitor I, CAS 1177749-58-4, is a cell-permeable, potent, and reversible MMP-9 Inhibitor (IC₅₀ = 5 nM). At high concentration inhibits MMP-1 (IC₅₀ = 1.05 µM) & MMP-13 (IC₅₀ = 113 nM).

Quality Level

Assay

≥93% (HPLC)

form

solid

potency

5 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

storage temp.

−20°C

General description

A cell-permeable, potent, selective, and reversible MMP-9 Inhibitor (IC50 = 5 nM). Inhibits MMP-1 (IC50 = 1.05 µM) and MMP-13 (IC50 = 113 nM) only at much higher concentrations.
A cell-permeable, potent, selective, and reversible inhibitor of MMP-9 (IC50 = 5 nM). Also inhibits MMP-1 (IC50 = 1.05 µM) and MMP-13 IC50 = 113 nM) at higher concentrations.

Biochem/physiol Actions

Primary Target
MMP-9

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Levin, J.I., et al. 2001. Bioorg. Med. Chem. Lett.11, 2189.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service