Skip to Content
Merck
All Photos(1)

Key Documents

444244

Sigma-Aldrich

MMP-2 Inhibitor I

The MMP-2 Inhibitor I, also referenced under CAS 10335-69-0, controls the biological activity of MMP-2. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2 Inhibitor I, cis-9-Octadecenoyl-N-hydroxylamide, Oleoyl-N-hydroxylamide, OA-Hy

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$199.00

$199.00


Estimated to ship on02 June 2025



Select a Size

Change View
10 MG
$199.00

About This Item

Empirical Formula (Hill Notation):
C18H35NO2
CAS Number:
Molecular Weight:
297.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$199.00


Estimated to ship on02 June 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

potency

1.7 mM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 2 mg/mL
ethanol: 3 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N(O)C(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C18H35NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(20)19-21/h9-10,21H,2-8,11-17H2,1H3,(H,19,20)/b10-9-

InChI key

LTXHELLMCCEDJG-KTKRTIGZSA-N

General description

A cell-permeable, potent inhibitor of MMP-2 that acts in a dose-dependent manner (Ki = 1.7 µM).
Potent inhibitor of MMP-2 (gelatinase A). Inhibits MMP-2 in a dose dependent manner (Ki = 1.7 µM)

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-2
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Berton, A., et. al. 2001. J. Biol. Chem.276, 20458.
Emonard, H., et al. 1999. Ann. N.Y. Acad. Sci.878, 647.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Hsi Chiu et al.
The FEBS journal, 281(10), 2353-2365 (2014-03-25)
Ste20-related proline/alanine-rich kinase (SPAK) plays a role in regulating many biological activities, and interacts with K-Cl co-transporter 3 (KCC3); however, the importance of SPAK for KCC3 function has not been demonstrated. Here, we investigated the role of SPAK in KCC3-regulated
Veronika Brezovakova et al.
Cells, 11(9) (2022-05-15)
Astrocytes regulate important functions in the brain, and their dysregulation has been linked to the etiology of neurodegenerative diseases, such as Alzheimer's disease (AD). The role of astroglia in human AD remains enigmatic, owing to the limitations of animal models

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service