Skip to Content
Merck
All Photos(3)

Key Documents

PLA0071

Sigma-Aldrich

Rabbit anti-Phospho RPA32 (S4/S8) Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

32kDa, REPA2, RF-A protein 2, RP-A p32, RP-A p34, RPA32, replication factor A protein 2, replication protein A 34 kDa subunit, replication protein A2, replication protein A2 (32kD)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$642.00

$642.00


Check Cart for Availability


Select a Size

Change View
100 μL
$642.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$642.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunocytochemistry: 1:500-1:5,000
immunohistochemistry: 1:500-1:2,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_002937.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer4/pSer8)

Gene Information

human ... RPA2(6118)

General description

Replication protein A2 (RPA32) is encoded by the gene mapped to human chromosome 1p35.3. It is a subunit of the heterotrimeric replication protein A (RPA) complex.

Immunogen

Immunogen was a dually phosphorylated synthetic peptide, which represented a portion of human replication protein A2, 32 kDa surrounding phosphorylated serines that corresponded to positions 4 and 8 using the numbering given in entry NP_002937.1 (GeneID 6118).

Biochem/physiol Actions

Replication Protein A (RPA) binds single-stranded DNA and plays a vital role in DNA replication, DNA repair, and checkpoint signaling. Phosphorylation of replication protein A2 (RPA32) by ataxia telangiectasia and Rad3-related protein kinase (ATR) and DNA-dependent protein kinase (DNA-PK) causes checkpoint kinase 1 (Chk1) activation and replication arrest. DNA-PK and ATR signaling via RPA32 (S4/S8) enhances genome stability and cell survival following replication stress.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Replication protein A (RPA) is a multisubunit complex that carries out DNA mismatch repair (MMR) in association with MSH2 (a subunit of human MutS heterodimers), MLH1 (a subunit of human MutL heterodimers), PCNA, and DNA polymerase-delta. RPA is composed of a heterotrimer that includes subunits of 70 kDa (RPA1), 32kDa (RPA2), and 14kDa (RPA3). RPA2, the 32kDa subunit, is phosphorylated by the cdc2 family of kinases when cells enter S-phase and in response to DNA damage by ATM, ATR, and DNA-PK.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial DNA repair genes and their eukaryotic homologues: 4. The role of nucleotide excision DNA repair (NER) system in mammalian cells.
Maddukuri L, et al.
Acta Biochimica Polonica, 54(3), 469-482 (2007)
DNA-PK phosphorylation of RPA32 Ser4/Ser8 regulates replication stress checkpoint activation, fork restart, homologous recombination and mitotic catastrophe
Ashley AK, et al.
DNA Repair, 21(3), 131-139 (2014)
Radka Storchova et al.
The FEBS journal, 288(20), 6035-6051 (2021-05-14)
Upon exposure to genotoxic stress, cells activate DNA damage response (DDR) that coordinates DNA repair with a temporal arrest in the cell cycle progression. DDR is triggered by activation of ataxia telangiectasia mutated/ataxia telangiectasia and Rad3-related protein kinases that phosphorylate
Alina Vaitsiankova et al.
Nature structural & molecular biology, 29(4), 329-338 (2022-03-26)
Poly(ADP-ribose) polymerase 1 (PARP1) is implicated in the detection and processing of unligated Okazaki fragments and other DNA replication intermediates, highlighting such structures as potential sources of genome breakage induced by PARP inhibition. Here, we show that PARP1 activity is
George Rasti et al.
Nucleic acids research, 51(13), 6754-6769 (2023-06-13)
The Sirtuin family of NAD+-dependent enzymes plays an important role in maintaining genome stability upon stress. Several mammalian Sirtuins have been linked directly or indirectly to the regulation of DNA damage during replication through Homologous recombination (HR). The role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service