Key Documents
PL800GB
Plas-Labs butyl glove box gloves
for 1 piece glove set, size 9 (for 8 in. ports)
Select a Size
1 PAIR
$1,180.00
$1,180.00
Available to ship on24 April 2025Details
Select a Size
Change View
1 PAIR
$1,180.00
About This Item
UNSPSC Code:
46181504
NACRES:
HA.03
$1,180.00
Available to ship on24 April 2025Details
material
butyl rubber
size
9 (for 8 in. ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GB
General description
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)