This product is intended for single use. Repeated usage may result in declining performance.
UFC5100
Amicon® Ultra Centrifugal Filter, 100 kDa MWCO
sample volume 0.5 mL, regenerated cellulose membrane, MWCO 100 kDa
Synonym(s):
Centrifugal concentrator, 100 kDa MWCO, 0.5 mL sample volume
Select a Size
Select a Size
About This Item
Recommended Products
material
polypropylene cap
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing
Quality Level
sterility
non-sterile
product line
Amicon®
packaging
pkg of 24 pkg
pkg of 500 pkg
pkg of 8 pkg
pkg of 96 pkg
manufacturer/tradename
Amicon® Ultra
sustainability
Greener Alternative Product
parameter
0.5 mL sample volume
technique(s)
protein extraction: suitable
protein purification: suitable
L
4.99 cm (1.96 in.)
diam.
10.8 mm
filtration area
1 cm2
volume
0.5 mL
working volume
≤500 μL
0.5 mL
pore size
100 kDa MWCO
greener alternative category
shipped in
ambient
General description
The vertical design and available membrane surface area provide fast sample processing, high sample recovery (typically greater than 90% of dilute starting solution), and the capability for 30-fold concentration. Solute polarization and subsequent fouling of the membrane are minimized by the vertical design, and a physical deadstop in the filter device prevents spinning to dryness and potential sample loss. Efficient recovery of the concentrated sample (retained species) is achieved by a convenient reverse spin step after collecting the filtrate.
Amicon® Ultra-0.5 devices are supplied non-sterile and are for single use only. With their vertical membrane design and reverse spin, Amicon® Ultra 0.5 mL filters deliver great performance and lower spin times.
Application
- Ultrafiltration with 100,000 Dalton molecular weight cutoff Ultracel® membrane
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates, primer removal, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
- Desalting, buffer exchange, and protein dialysis
Features and Benefits
- Ultracel® regenerated cellulose membrane with 100 kDa NMWCO delivers retentate recovery of greater than 90%
- Concentrates 500 μL down to 15 μL
- Concentration factor 25x to 30x
- High sample recovery (greater than 90% of dilute starting solution)
- Fast processing time: 10 to 30 minutes
- Reverse spin capability provides consistent recoveries
Linkage
Physical form
Legal Information
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Commercially available protein labeling kits provide researchers with the flexibility to customize their own immunoassay detection panels.
Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.
Filtration methods and membrane filter selection in nanoparticle purification and production.
Explore ultrafiltration membrane principles and benefits for protein sample preparation, including solvent exchange and sample concentration.
Protocols
If an antibody must be purified from a small sample volume (0.5-10 mL), then the Amicon® Pro Purification System is recommended.
Detergents are frequently used to solubilize proteins and nucleic acids during purification, but the presence of detergents may interfere with downstream analyses.
-
Can I reuse Amicon® Ultra Centrifugal Filter, 100 kDa MWCO? ?
1 answer-
Helpful?
-
-
Que signifie le conditionnement de "8 pkg"? Cela correspond-il à 8 eppendorfs/filtres ?
1 answer-
Yes, "8 pkg" means a package of 8 Amicon® Ultra Centrifugal Filters.
Helpful?
-
-
Is dimethylformamide chemically compatible with Amicon Ultra-0.5mL?
1 answer-
No data is presently available for compatibility with dimethylformamide.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service