Skip to Content
Merck
All Photos(1)

Key Documents

MBD6009

Sigma-Aldrich

5R-PLEX beads

for PCR purification

Synonym(s):

5R-PLEX SPRI beads

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$251.00

$251.00


Available to ship on14 April 2025Details



Select a Size

Change View
5 ML
$251.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.55

$251.00


Available to ship on14 April 2025Details


Quality Level

form

suspension

technique(s)

PCR: suitable

storage temp.

2-8°C

General description

Solid Phase Reversible Immobilization (SPRI) beads are magnetic beads that can be utilized for DNA purification and size selection. The 5R-PLEX beads are SPRI beads designed to easily separate the desired PCR amplicon from nucleotides, adapters, primers, and primer dimers by efficient purification and size selection. The magnetic beads are provided in a ready to use suspension that enables fast and efficient application even in high throughput applications with automated liquid handling. The beads are formulated with NaCl and high purity PEG as the crowding agent.

Application

  • NGS library preparation, including 16S and 18S
  • Approved to use in ultrasensitive 5R-Plex (SMURF) library preparation(2,3,4) for degraded samples
  • PCR reaction clean up
  • Amplicon size selection
  • DNA purification and manipulation (replacing elution buffer, concentrating the sample)

Features and Benefits

  • Ready to use
  • Compatible with automated liquid handling
  • Easy size selection by changing the volumetric ratio of beads to samples

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lishay Parhi et al.
Nature communications, 11(1), 3259-3259 (2020-06-28)
Fusobacterium nucleatum is an oral anaerobe recently found to be prevalent in human colorectal cancer (CRC) where it is associated with poor treatment outcome. In mice, hematogenous F. nucleatum can colonize CRC tissue using its lectin Fap2, which attaches to
Solid-phase reversible immobilization for the isolation of PCR products.
M M DeAngelis et al.
Nucleic acids research, 23(22), 4742-4743 (1995-11-25)
[Studies on 7 patients with giardiasis].
K Iguchi et al.
Rinsho byori. The Japanese journal of clinical pathology, 36(10), 1223-1226 (1988-10-01)
Archita Mishra et al.
Cell, 184(13), 3394-3409 (2021-06-03)
The human fetal immune system begins to develop early during gestation; however, factors responsible for fetal immune-priming remain elusive. We explored potential exposure to microbial agents in utero and their contribution toward activation of memory T cells in fetal tissues. We
Yunuen Avalos-Padilla et al.
iScience, 26(1), 105765-105765 (2023-01-03)
The endosomal sorting complex required for transport (ESCRT) is a multi-protein machinery involved in several membrane remodeling processes. Different approaches have been used to resolve how ESCRT proteins scission membranes. However, the underlying mechanisms generating membrane deformations are still a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service