Skip to Content
Merck
All Photos(2)

Key Documents

GE29357373

SeraSil-Mag 700 silica coated superparamagnetic beads

Cytiva 29357373, pack of 1 × 5 mL

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$155.00

$155.00


Available to ship on28 April 2025Details



Select a Size

Change View
5 ML
$155.00

About This Item

UNSPSC Code:
41105507
NACRES:
NB.13

$155.00


Available to ship on28 April 2025Details


packaging

pack of 1 × 5 mL

manufacturer/tradename

Cytiva 29357373

avg. part. size

400 nm

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

SeraSil-Mag silica coated superparamagnetic beads for nucleic acid isolation deliver high purity DNA extraction for highly sensitive applications where sample is scarce. These beads provide an optimal surface for nucleic acid binding with high performance and low background. For ongoing reliability,SeraSil-Mag is supported by Cytiva’s expertise in Sera-Mag bead technology and commitment to quality standards.

SeraSil-Mag beads are compatible with chaotropic salt chemistry and vortex mixing, roller mixing and sonication.

Tightly controlled production processes result in core-shell beads which are uniform and spherical. The silica shell provides a high content of silanol surface groups and encloses the magnetic core. The submicron size allows for a high surface area and abundant binding sites. The narrow particle size distribution assures uniformity and consistency of the beads.

The magnetic nanoparticles within the core provide superparamagnetic properties to the SeraSil-Mag beads, resulting in extremely fast magnetic separation in the presence of a magnet. There is no residual magnetization when the magnetic field is removed, which further contributes to ease of handling during use.

Features and Benefits

  • High magnetization (60emu/g) and strong binding capacity giving Fast magnetic response (~5 secs) and shorten time required for magnetic steps during isolation.
  • Beads are monodispersed with narrow size distribution for consistent, reproducible results.
  • Silanol hydroxyl groups on the bead surface give efficient, high purity isolation of nucleic acids.
  • Tested for microbial contamination.
  • Good buoyancy stability (low sedimentation rate): beads remain well suspended and dispersed for over 90 mins (at 1 mg/ml concentration),* providing good handling and suitability for automation.
  • Beads may settle faster at higher concentration
  • Beads come in two sizes (submicroscale diameter ~700 nm and 400 nm), offering choice based on surface requirements.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service