Skip to Content
Merck
All Photos(1)

Key Documents

M0383

Sigma-Aldrich

Malt extract

from starch digestion

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
eCl@ss:
42040102

biological source

plant (Hordeum vulgare)

form

powder

Looking for similar products? Visit Product Comparison Guide

General description

Malt extract represents the soluble material extracted from malt during the mashing process of barley.

Application

A source of carbohydrates.
Petri dishes filled with malt extract agar, separately inoculated with fungal strains and cultivated to obtain an abundant and even growth, were used as the test surface in a study assessing the effect of two aerosolization methods on the release of fungal propagules from a contaminated agar surface. Malt extract has also been used in a kinetic study to investigate microbial growth using the Monod equation.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mapping and validation of chromosome regions associated with high malt extract in barley (Hordeum vulgare L.)
Collins, H., et al.
Australian Journal of Agricultural Research, 54, 1223-1240 (2003)
Rafał L Górny et al.
Annals of agricultural and environmental medicine : AAEM, 19(2), 279-284 (2012-06-30)
The effect of perpendicular and swirling aerozolization methods on the release of fungal fragments and spores from agar surface was studied. Three fungal species (Aspergillus versicolor, Cladosporium cladosporioides, Penicillium chrysogenum) were selected for the tests as they commonly occur indoors
Ajeng Arum Sari et al.
Journal of bioscience and bioengineering, 114(2), 176-181 (2012-05-18)
Trametes versicolor U97 isolated from nature degraded 73% of the 1,1,1-trichloro-2,2-bis(4-chlorophenyl) ethane (DDT) in a malt extract liquid medium after a 40-d incubation period. This paper presents a kinetic study of microbial growth using the Monod equation. T. versicolor U97

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service