Skip to Content
Merck
All Photos(1)

Key Documents

M6409

Millipore

Malt Extract Broth

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$166.00

$166.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$166.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

$166.00


Available to ship on07 April 2025Details


Request a Bulk Order

description

Microbiologically tested

Quality Level

sterility

non-sterile

form

powder

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

application(s)

bioburden testing
food and beverages

microbiology

suitability

molds
yeasts

Application

Recommended for use in the cultivation of recombinant strains of E. coli.

Components

Contains: Malt extract, maltose, yeast extract and dextrose.

Preparation Note

Solve 15.0 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service