Merck
  • Determination of co-metabolism for 1,1,1-trichloro-2,2-bis(4-chlorophenyl) ethane (DDT) degradation with enzymes from Trametes versicolor U97.
Determination of co-metabolism for 1,1,1-trichloro-2,2-bis(4-chlorophenyl) ethane (DDT) degradation with enzymes from Trametes versicolor U97.
Journal of bioscience and bioengineering (2012-05-18)
Ajeng Arum Sari, Sanro Tachibana, Kazutaka Itoh
ABSTRACT
Trametes versicolor U97 isolated from nature degraded 73% of the 1,1,1-trichloro-2,2-bis(4-chlorophenyl) ethane (DDT) in a malt extract liquid medium after a 40-d incubation period. This paper presents a kinetic study of microbial growth using the Monod equation. T. versicolor U97 degraded DDT during an exponential growth phase, using glucose as a carbon source for growth. The growth of T. versicolor U97 was not affected by DDT. DDT was degraded by T. versicolor U97 only when the secondary metabolism coincided with the production of several enzymes. Furthermore, modeling of several inhibitors using the partial least squares function in Minitab 15, revealed lignin peroxidase (98.7 U/l) plays a role in the degradation of DDT. T. versicolor U97 produced several metabolites included a single-ring aromatic compound, 4-chlorobenzoic acid.
Sigma-Aldrich
4-Chlorobenzoic acid, 99%
Sigma-Aldrich
Ethane, 99.99%
Sigma-Aldrich
Ethane, ≥99%