Skip to Content
Merck

Skip To

L1663

Lidocaine N-ethyl chloride

sodium channel blocker

Synonym(s):

N-(2,6-Dimethylphenylcarbamoylmethyl) triethylammonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 mg
Check Cart for Availability
$573.00
1 g
Check Cart for Availability
$836.00
$710.60

About This Item

Empirical Formula (Hill Notation):
C16H27N2OCl
CAS Number:
Molecular Weight:
298.85
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Quality level:
Storage condition:
protect from light

$573.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Lidocaine N-ethyl chloride,

storage condition

protect from light

Quality Level

solubility

H2O: soluble

storage temp.

room temp

SMILES string

[Cl-].CC[N+](CC)(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C16H26N2O.ClH/c1-6-18(7-2,8-3)12-15(19)17-16-13(4)10-9-11-14(16)5;/h9-11H,6-8,12H2,1-5H3;1H

InChI key

LLPPOMUAOGMYQI-UHFFFAOYSA-N

Biochem/physiol Actions

Lidocaine N-ethyl chloride is an intracellular voltage-gated sodium channel blocker.

Other Notes

Stable at room temperature lyophilized. In solution, store at 4 °C for up to two weeks. Store solutions at -20 °C for up to six months.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1444C4382D2926
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

300

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage condition

protect from light

storage condition

desiccated

storage condition

-

storage condition

-

solubility

H2O: soluble

solubility

H2O: 20 mg/mL, clear

solubility

H2O: 1 g/mL, ethanol: 20 mg/mL

solubility

DMSO: 9.80-10.20 mg/mL, clear, colorless to faintly yellow


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Vijay K Samineni et al.
Nature communications, 10(1), 4356-4356 (2019-09-27)
Itch is a distinct aversive sensation that elicits a strong urge to scratch. Despite recent advances in our understanding of the peripheral basis of itch, we know very little regarding how central neural circuits modulate acute and chronic itch processing.
Iina Korkka et al.
Stem cells translational medicine, 8(2), 179-193 (2018-11-06)
Retinal pigment epithelium (RPE) performs important functions for the maintenance of photoreceptors and vision. Malfunctions within the RPE are implicated in several retinal diseases for which transplantations of stem cell-derived RPE are promising treatment options. Their success, however, is largely
Andrew W Baxter et al.
The European journal of neuroscience, 34(2), 213-220 (2011-07-14)
P2X4 receptors are calcium-permeable cation channels gated by extracellular ATP. They are found close to subsynaptic sites on hippocampal CA1 neurons. We compared features of synaptic strengthening between wild-type and P2X4 knockout mice (21-26 days old). Potentiation evoked by a



Global Trade Item Number

SKUGTIN
L1663-500MG04061832574783
L1663-1G04061826226247

Questions

Reviews

No rating value

Active Filters