Merck
HomeProductsPharma & Biopharma ManufacturingChromatographyChromatography Columns for Bioprocessing
Chromatography Columns for Bioprocessing
Chromatography columns are critical to the successful separation of your valuable molecules. We provide columns from lab scale to pilot and process scale, delivering robust and consistent performance with the processing flexibility you need for anything from screening to large-scale production.
Read more about
Products
Sort by Relevance
Showing 1-1 of 1
Change View
Showing 1-1 of 1
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-1 of 1 results
Page 1 of 1
Page 1 of 1
Examples of the various sizes of Vantage® chromatography columns
Vantage® columns for research, development, and production
Vantage® chromatography columns are ideal for development, pilot, or production purification in pharmaceutical and biotechnology applications. Vantage® columns are designed for simple assembly, scalability from R&D to pilot at small-scale production, and zero dead space.
  • Our Vantage® L family of laboratory columns for research and development are ideally suited for small-particle media applications
  • Our Vantage® A2 family of chromatography columns are well-suited for rigid, higher-resolution chromatography media
  • Our Vantage® S2 chromatography columns provide flexibility for a wide range of applications
An example of a QuikScale® chromatography columns, includes a clear glass bottle attached to a stand, set on a white background. The stand has four metal legs with wheels for mobility
QuikScale® scalable family of columns
This robust, scalable family of chromatography columns maximizes process yields, boosts productivity, and shortens time to market. QuikScale® columns achieve ultra-high throughput and deliver greater product purity – at faster linear velocities. They accommodate all media types, are easily and uniformly packed, and deliver optimal resolution across a wide range of applications.
An innovative flow distributor ensures full, uniform media utilization for reproducible and reliable separations. A high-flow option (with operating pressures up to 7 bar and flow rates up to 1,000 cm/hr) maximizes process design flexibility. This allows greater bed height settings when handling media with high flow and/or back pressure requirements.
QuikScale® chromatography columns are designed for:
  • Scalability with a full range of column diameters (from 70 mm to 630 mm) and heights (550 mm to 1,100 mm), as well as bed support pore size (10, 20, and 30 µm)
  • Wide variety of applications – thanks to multiple tube materials (glass, acrylic, stainless steel)
Stainless steel circular chromatography column mounted on a stand. It has multiple vertical tubes connected by metal rings at the top and bottom. Adjustment knobs and levers are present on the stand below the column. The equipment is mounted on a metal stand with caster wheels for mobility.
IsoPak™ columns for process-scale applications
IsoPak™ columns are fast and highly reproducible, delivering consistent chromatographic results that are ideal for process-scale applications. What sets an IsoPak™ column apart is its reproducible packing and unpacking. Due to a unique valve design, slurry is pumped directly into the column, enabling the column to be packed and unpacked without the removal of the top flow cell. By enclosing operations, exposure between the operator and the chromatographic media is drastically limited.
IsoPak™ columns are available in a wide range of diameters (440 to 1,400 mm and higher), tubing materials, and bed heights (500 to 1,000 mm). They provide:
  • Consistent packing and unpacking for reproducible results
  • Ease of maintenance
  • Scalability
2 different sizes of minichrom prepacked chromatography columns, designed for process development
Minichrom Columns for Process Development
MiniChrom columns are designed for process development, method optimization, and parameter screening as well as sample preparation. MiniChrom columns are quality checked for HETP and asymmetry, providing:
  • Superior reproducibility
  • Excellent scalability
  • Convenient comparison of different chromatography media
8 robocolumns prepacked chromatography columns arranged on a microplate
Robocolumns® for High Throughput Processes
RoboColumns® are easy-to-use, miniaturized chromatography columns which can be individually arranged on the commonly used and standard 96 well microplate format according to application requirements. They are designed for high throughput process development, resin and condition screening and optimization, and sample preparation prior to subsequent analysis.
1 Hibar® prepacked chromatograpghy columns, made of stainless steel
Hibar® Columns for Media Screening
Ready-to-use stainless steel columns prepacked with chromatography sorbents for the separation of mg up to grams range of final product.
9 different sizes of Opus® prepacked chromatography columns, aligned, from the smallest size on the left to the biggest size on the right
Opus® Columns for Scalability and Flexibility
From benchtop to production, our full portfolio of resins is now available in a single-use, plug and play format. The collaborative offering combines our high performance resins with the industry leading Repligen’s OPUS® pre-packed columns for ultimate flexibility. OPUS® columns provide linear performance scale up from process validation to manufacturing.
Related Product Resources
Page 1 of 3