L0277
Cryoware labels, plain
sheet of 20 labels
Synonym(s):
cold storage labels, cryo labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Recommended Products
material
fabric
white
packaging
pkg of 10 sheets
sheet of 20 labels
manufacturer/tradename
Nalgene® 5040-0002
technique(s)
cryopreservation: suitable
L × W
25 mm × 50 mm
Related Categories
General description
Durable cloth labels, 1 × 2 in. (25 × 50 mm) for use at ultra-low temperature. Adhere to plastic and cardboard storage boxes without peeling or shrinking.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service