Skip to Content
Merck
All Photos(1)

Documents

Z359017

Nalgene® CryoBox

CryoBox 100, 10 × 10 array

Synonym(s):

cryovial storage box, freezer storage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.22

material

polycarbonate

feature

Holds 100 x 1.2 -2.0 mL vials

packaging

pack of 10 ea

manufacturer/tradename

Nalgene® 5026-1010

technique(s)

cryopreservation: suitable

L × W × H

5 1/4 in. × 5 1/4 in. × 2 1/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

For ultra-compact storage of 1.0ml and 1.5ml System 100 vials or other similar-sized vials. Box fits most existing vertical racks with 2in. shelf spacing.
For ultra-low temperature storage of cryogenic vials in mechanical or liquid nitrogen freezers. Usable temperature range: −196 to +121 °C. Numbers molded into grid system in box correspond to numbers printed on lid. Lid is keyed to box so markings will always match. Boxes accept writing with markers.

Legal Information

CryoBoxes is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service